missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-12/IL-23 Mouse anti-Human, eFluor 450, Clone: HP40, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen IL-12/IL-23
Clone HP-40)
Host Species Murine
Gene Alias Interleukin-12/ Interleukin-23
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124213 Life Technologies
LS48723541
25 Tests Each for $143.55
501124214 Life Technologies
LS48723542
100 Tests Each for $361.05
Specifications

Description

The eBioHP40 antibody reacts with the p40 subunit of human IL-12 p70, and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.

The eBioHP40 antibody has slightly lower background staining than C8.6 and recognizes a different epitope.
Description & Specifications

Specifications

IL-12/IL-23
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HP-40)
Interleukin-12/ Interleukin-23
Flow Cytometry (Intracellular Staining)
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test