Please login to your online account to display your discounted pricing

IL-12/IL-23 Mouse anti-Human, Clone: C8.6, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$129.05 - $404.55

Specifications

Antigen IL-12/IL-23
Clone C8.6
Host Species Mouse
Gene Alias Interleukin-12/Interleukin-23, p40/70
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122986

 
life technologies
LS16712981
50μg Each for $129.05

501122987

 
life technologies
LS16712985
500μg Each for $404.55
Description & Specifications

Specifications

Antigen IL-12/IL-23
Clone C8.6
Host Species Mouse
Gene Alias Interleukin-12/Interleukin-23, p40/70
Species Reactivity Human
Applications Cytokine Neutralization
Applications ELISPOT
Applications Enzyme Linked Immunosorbent Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.