missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-10 Rat anti-Mouse, PerCP-Cy5.5, Clone: JES5-16E3, eBioscience™

Rat Monoclonal Antibody

$191.40 - $442.25

Specifications

Antigen IL-10
Clone JES5-16E3
Host Species Rat
Gene Alias Interleukin-10
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015871 Life Technologies
LS45710180
25μg Each for $191.40
5015872 Life Technologies
LS45710182
100μg Each for $442.25
Specifications

Description

The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). Mouse IL-10 is an ∽18kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.
Description & Specifications

Specifications

IL-10
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
JES5-16E3
Interleukin-10
Flow Cytometry (Intracellular Staining)
PerCP-Cy5.5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL