Please login to your online account to display your discounted pricing

IL-10 Rat anti-Mouse, FITC, Clone: JES5-16E3, eBioscience™

Rat Monoclonal Antibody

$114.55 - $274.05

Specifications

Antigen IL-10
Clone JES5-16E3
Host Species Rat
Gene Alias Interleukin-10, IL10
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010009

 
life technologies
LS11710182
100μg Each for $274.05

5010008

 
life technologies
LS11710181
50μg Each for $143.55

501122153

 
life technologies
LS11710141
25 Tests Each for $114.55
Description & Specifications

Specifications

Antigen IL-10
Clone JES5-16E3
Host Species Rat
Gene Alias Interleukin-10, IL10
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL;, 5μL (0.25μg)/test

The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). Mouse IL-10 is an ∽18kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.