Please login to your online account to display your discounted pricing

IL-10 Rat anti-Mouse, Clone: JES5-2A5, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$143.55 - $419.05

Specifications

Antigen IL-10
Clone JES5-2A5
Host Species Rat
Gene Alias Interleukin-10, IL10
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014714

 
life technologies
LS16710281
50μg Each for $143.55

5014715

 
life technologies
LS16710285
500μg Each for $419.05
Description & Specifications

Specifications

Antigen IL-10
Clone JES5-2A5
Host Species Rat
Gene Alias Interleukin-10, IL10
Species Reactivity Mouse
Applications Cytokine Neutralization
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The JES5-2A5 antibody reacts with mouse interleukin-10 (IL-10). The JES5-2A5 antibody is a neutralizing antibody. Mouse IL-10 is an 18kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.