missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-10 Rat anti-Mouse, Biotin, Clone: JES5-2A5, eBioscience™

Rat Monoclonal Antibody

$130.68 - $341.88

Specifications

Antigen IL-10
Clone JES5-2A5
Host Species Rat
Gene Alias Interleukin-10
Applications ELISA Detection,ELISPOT Detection Antibody
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012170 Life Technologies
LS13710285
500μg Each for $341.88
501128836 Life Technologies
LS13710281
50μg Each for $130.68
Specifications

Description

The JES5-2A5 antibody reacts with mouse interleukin-10 (IL-10). The JES5-2A5 antibody is a neutralizing antibody. Mouse IL-10 is an 18kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.
Description & Specifications

Specifications

IL-10
Rat
ELISA Detection,ELISPOT Detection Antibody
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
JES5-2A5
Interleukin-10
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer