missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-10, APC, clone: JES5-16E3, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS17710181

 View more versions of this product

Catalog No. 501129673



Description

Description

Interleukin-10, originally known as Cytokine Synthesis Inhibitory Factor (CSIF), is an 18.6kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein BCRFI. The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production.

Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.
Specifications

Specifications

IL-10
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P18893
Interleukin-10, CSIF, IL-10, IL10A, TGIF
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
JES5-16E3
APC
Il10
Liquid
Il10
IgG2b, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16153
Documents

For Research Use Only.