missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-10 Rat anti-Human, PE-Cy7, Clone: JES3-9D7, eBioscience™

Rat Monoclonal Antibody

$174.24 - $443.52

Specifications

Antigen IL-10
Clone JES3-9D7
Host Species Rat
Gene Alias Interleukin-10
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123434 Life Technologies
LS25710841
25 Tests Each for $174.24
501129197 Life Technologies
LS25710842
100 Tests Each for $443.52
Specifications

Description

The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10).
Description & Specifications

Specifications

IL-10
Rat
Flow Cytometry (Intracellular Staining)
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.06μg)/test
JES3-9D7
Interleukin-10
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer