Please login to your online account to display your discounted pricing

IL-10 Rat anti-Human, PE, Clone: JES3-9D7, eBioscience™

Rat Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen IL-10
Clone JES3-9D7
Host Species Rat
Gene Alias Interleukin-10
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011149

 
affymetrix
12710841
25 tests Each for $143.55

5011150

 
affymetrix
12710881
50μg Each for $201.55

5011151

 
affymetrix
12710882
100μg Each for $361.05
Description & Specifications

Specifications

Antigen IL-10
Clone JES3-9D7
Host Species Rat
Gene Alias Interleukin-10
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.125μg)/test

The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10).