Please login to your online account to display your discounted pricing

IL-10 Rat anti-Human, Clone: JES3-9D7, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$143.55 - $2,463.55

Specifications

Antigen IL-10
Clone JES3-9D7
Host Species Rat
Gene Alias Interleukin-10
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122984

 
affymetrix
16710838
5mg Each for $2,463.55

5014716

 
affymetrix
16710881
50μg Each for $143.55

501129462

 
affymetrix
16710885
500μg Each for $419.05
Description & Specifications

Specifications

Antigen IL-10
Clone JES3-9D7
Host Species Rat
Gene Alias Interleukin-10
Species Reactivity Human
Applications Cytokine Neutralization
Applications ELISPOT Capture Antibody
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10).