missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-10 Rat anti-Human, Biotin, Clone: JES3-12G8, eBioscience™

Rat Monoclonal Antibody

$143.55 - $375.55

Specifications

Antigen IL-10
Clone JES3-12G8
Host Species Rat
Gene Alias Interleukin-10, IL10
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012171 Life Technologies
LS13710981
50μg Each for $143.55
5012172 Life Technologies
LS13710985
500μg Each for $375.55
Specifications

Description

The JES3-12G8 antibody reacts with human interleukin-10 (IL-10).
Description & Specifications

Specifications

IL-10
Rat
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
JES3-12G8
Interleukin-10, IL10
ELISA Detection,ELISPOT Detection Antibody
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL