Please login to your online account to display your discounted pricing

IL-10 Rat anti-Human, Alexa Fluor 488, Clone: JES3-9D7, eBioscience™

Rat Monoclonal Antibody

$172.55 - $361.05

Specifications

Antigen IL-10
Clone JES3-9D7
Host Species Rat
Gene Alias Interleukin-10
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016796

 
affymetrix
53710841
25 tests Each for $172.55

5016797

 
affymetrix
53710842
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen IL-10
Clone JES3-9D7
Host Species Rat
Gene Alias Interleukin-10
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10).