missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-1 beta Mouse anti-Human, FITC, Clone: CRM56, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $289.08

Specifications

Antigen IL-1 beta
Clone CRM56
Host Species Murine
Gene Alias Interleukin-1 beta
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509986 Life Technologies
LS11701841
25 Tests Each for $117.48
509987 Life Technologies
LS11701842
100 Tests Each for $289.08
Specifications

Description

The CRM56 antibody reacts with human and baboon interleukin-1 beta; (IL-1 beta). IL-1 beta is a 17kDa factor secreted primarily by monocytes. IL-1 has effects on T-helper cells, inducing IL-2 secretion and expression of IL-2 receptors. IL-1 has effects on B cells, promoting cell proliferation and immunoglobulin synthesis.
Description & Specifications

Specifications

IL-1 beta
Murine
Flow Cytometry (Intracellular Staining)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
CRM56
Interleukin-1 beta
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer