missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-1 beta Mouse anti-Human, Clone: CRM56, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$130.68 - $368.28

Specifications

Antigen IL-1 beta
Clone CRM56
Host Species Murine
Gene Alias Interleukin-1 beta
Applications Cytokine Neutralization,ELISA Capture,ELISPOT
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014689 Life Technologies
LS16701881
50μg Each for $130.68
5014690 Life Technologies
LS16701885
500μg Each for $368.28
Specifications

Description

The CRM56 antibody reacts with human and baboon interleukin-1 beta; (IL-1 beta). IL-1 beta is a 17kDa factor secreted primarily by monocytes. IL-1 has effects on T-helper cells, inducing IL-2 secretion and expression of IL-2 receptors. IL-1 has effects on B cells, promoting cell proliferation and immunoglobulin synthesis.
Description & Specifications

Specifications

IL-1 beta
Murine
Cytokine Neutralization,ELISA Capture,ELISPOT
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
CRM56
Interleukin-1 beta
RUO
Purified
Primary
aqueous buffer, no sodium azide