Please login to your online account to display your discounted pricing

IL-1 beta Mouse anti-Human, Clone: CRM56, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $332.05

Specifications

Antigen IL-1 beta
Clone CRM56
Host Species Mouse
Gene Alias Interleukin-1 beta
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122722

 
life technologies
LS14701885
500μg Each for $332.05

5013592

 
life technologies
LS14701881
50μg Each for $129.05
Description & Specifications

Specifications

Antigen IL-1 beta
Clone CRM56
Host Species Mouse
Gene Alias Interleukin-1 beta
Species Reactivity Human
Applications Cytokine Neutralization
Applications ELISA Capture
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The CRM56 antibody reacts with human and baboon interleukin-1 beta; (IL-1 beta). IL-1 beta is a 17kDa factor secreted primarily by monocytes. IL-1 has effects on T-helper cells, inducing IL-2 secretion and expression of IL-2 receptors. IL-1 has effects on B cells, promoting cell proliferation and immunoglobulin synthesis.