Please login to your online account to display your discounted pricing

IL-1 alpha Mouse anti-Human, FITC, Clone: 364/3B3-14, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $346.55

Specifications

Antigen IL-1 alpha
Clone 364/3B3-14
Host Species Mouse
Gene Alias Interleukin-1 alpha
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010013

 
life technologies
LS11711881
50μg Each for $172.55

5010014

 
life technologies
LS11711882
100μg Each for $346.55
Description & Specifications

Specifications

Antigen IL-1 alpha
Clone 364/3B3-14
Host Species Mouse
Gene Alias Interleukin-1 alpha
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL; 5μL (0.5μg)/test

The 364/3B3-14 antibody reacts with human interleukin-1alpha.