missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IKBKB (IKK Beta) Recombinant Human Protein DFS Item

IKBKB (IKK Beta) Recombinant Human Protein

Manufacturer:  Life Technologies LSPV3836

 View more versions of this product

Catalog No. PV3836



Description

Description

Phosphorylation of serine residues on the I-kappa-B proteins by IKK kinases, including IKBKB, marks them for destruction via the ubiquitination pathway, allowing activation of the NF-kappa-B complex.
Specifications

Specifications

NP_001547
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
IKBKB
Baculovirus
Full Length, Recombinant
Signal Transduction
Kinase Assay
114 kDa
IKBKB (IKK Beta)
GST Tag
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Ser⁄Thr Kinases: Other
LanthaScreen® Antibody, LanthaScreen® Substrate, Z'-LYTE™, Omnia® Bulk Substrate
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
Kinases & Inhibitors
Serine⁄Threonine Kinases
Human
3551, 4088
10μg
IKK Kinase Family
Documents

For Research Use Only. Not for use in diagnostic procedures.