Please login to your online account to display your discounted pricing

IgM Rat, PE, Monoclonal, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$143.55 - $216.05

Specifications

Antigen IgM
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122313

 
life technologies
LS12434181
50μg Each for $143.55

5018649

 
life technologies
LS12434182
100μg Each for $216.05
Description & Specifications

Specifications

Antigen IgM
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal rat IgM, κ is useful as an isotype control immunoglobulin.