Please login to your online account to display your discounted pricing

IgM Rat, PE-eFluor 610, Monoclonal, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$129.05 - $317.55

Specifications

Antigen IgM
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124817

 
life technologies
LS61434182
100μg Each for $317.55

501124816

 
life technologies
LS61434180
25μg Each for $129.05
Description & Specifications

Specifications

Antigen IgM
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal rat IgM, κ is useful as an isotype control immunoglobulin.