Please login to your online account to display your discounted pricing

IgM Rat, PE-eFluor 610, Clone: II/41, eBioscience™

Rat Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen IgM
Clone II/41
Host Species Rat
Gene Alias Immunoglobulin M
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124839

 
life technologies
LS61579080
25μg Each for $158.05

501124840

 
life technologies
LS61579082
100μg Each for $390.05
Description & Specifications

Specifications

Antigen IgM
Clone II/41
Host Species Rat
Gene Alias Immunoglobulin M
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The II/41 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated II/41 can be used as a detection secondary for mouse IgM.