Please login to your online account to display your discounted pricing

IgM Rat, Monoclonal, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$114.55 - $172.55

Specifications

Antigen IgM
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013042

 
life technologies
LS14434185
500μg Each for $172.55

5013041

 
life technologies
LS14434182
100μg Each for $114.55
Description & Specifications

Specifications

Antigen IgM
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal rat IgM, κ is useful as an isotype control immunoglobulin.