Please login to your online account to display your discounted pricing

IgM Rat anti-Mouse, PE, Clone: eB121-15F9, eBioscience™

Rat Monoclonal Antibody

$143.55 - $419.05

Specifications

Antigen IgM
Clone eB121-15F9
Host Species Rat
Gene Alias Immunoglobulin M, Ig M
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010957

 
life technologies
LS12589081
50μg Each for $143.55

5010958

 
life technologies
LS12589082
100μg Each for $274.05

5010959

 
life technologies
LS12589083
200μg Each for $419.05
Description & Specifications

Specifications

Antigen IgM
Clone eB121-15F9
Host Species Rat
Gene Alias Immunoglobulin M, Ig M
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eB121-15F9 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated eB121-15F9 can be used as a detection secondary for mouse IgM.