missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgM Rat anti-Mouse, eFluor 450, Clone: II/41, eBioscience™

Rat Monoclonal Antibody

$121.44 - $299.64

Specifications

Antigen IgM
Clone II/41
Host Species Rat
Gene Alias Immunoglobulin M
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124173 Life Technologies
LS48579080
25μg Each for $121.44
501128647 Life Technologies
LS48579082
100μg Each for $299.64
Specifications

Description

The II/41 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated II/41 can be used as a detection secondary for mouse IgM.
Description & Specifications

Specifications

IgM
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
II/41
Immunoglobulin M
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL