missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgM Rat anti-Mouse, Biotin, Clone: eB121-15F9, eBioscience™

Rat Monoclonal Antibody

$91.08 - $157.08

Specifications

Antigen IgM
Clone eB121-15F9
Host Species Rat
Gene Alias Immunoglobulin M, Ig M
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012001 Life Technologies
LS13589081
50μg Each for $91.08
5012002 Life Technologies
LS13589082
100μg Each for $157.08
Specifications

Description

The eB121-15F9 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated eB121-15F9 can be used as a detection secondary for mouse IgM.
Description & Specifications

Specifications

IgM
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eB121-15F9
Immunoglobulin M, Ig M
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer