Please login to your online account to display your discounted pricing

IgM Mouse anti-Human, eFluor 450, Clone: SA-DA4, eBioscience™

Mouse Monoclonal Antibody

$181.25 - $448.05

Specifications

Antigen IgM
Clone SA-DA4
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124243

 
life technologies
LS48999841
25 tests Each for $181.25

501124244

 
life technologies
LS48999842
100 tests Each for $448.05
Description & Specifications

Specifications

Antigen IgM
Clone SA-DA4
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The SA-DA4 monoclonal antibody reacts with the mu heavy chain of human IgM. It does not react with other classes of human immunoglobulin including IgD, IgG, or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of mature B cells.