Please login to your online account to display your discounted pricing

IgM Mouse anti-Human, Biotin, Clone: SA-DA4, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $158.05

Specifications

Antigen IgM
Clone SA-DA4
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012319

 
life technologies
LS13999880
25μg Each for $85.55

5012320

 
life technologies
LS13999882
100μg Each for $158.05
Description & Specifications

Specifications

Antigen IgM
Clone SA-DA4
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The SA-DA4 monoclonal antibody reacts with the mu heavy chain of human IgM. It does not react with other classes of human immunoglobulin including IgD, IgG, or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of mature B cells.

Our testing has revealed variations in staining of the non-B cell population that is predicted to be non-specific. This variation can be attributed to both donor variability and the anticoagulant used during blood draw. Therefore, we recommend using sodium citrate as the anticoagulant to minimize non-specific staining by this antibody.