missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgM Mouse anti-Human, APC, Clone: SA-DA4, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $376.20

Specifications

Antigen IgM
Clone SA-DA4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry,Immunohistochemistry (Frozen)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245717 Life Technologies
LS17999841
25 Tests Each for $170.28
50245718 Life Technologies
LS17999842
100 Tests Each for $376.20
Specifications

Description

The SA-DA4 monoclonal antibody reacts with the mu heavy chain of human IgM. It does not react with other classes of human immunoglobulin including IgD, IgG, or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of mature B cells.

Our testing has revealed variations in staining of the non-B cell population that is predicted to be non-specific. This variation can be attributed to both donor variability and the anticoagulant used during blood draw. Therefore, we recommend using sodium citrate as the anticoagulant to minimize non-specific staining by this antibody.
Description & Specifications

Specifications

IgM
Murine
Flow Cytometry,Immunohistochemistry (Frozen)
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
SA-DA4
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer