missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
IGHA1 Mouse anti-Human, Monkey, Mouse, Clone:7D5F12, Abnova™
Mouse monoclonal antibody raised against recombinant human IGHA1.
Supplier: Abnova Corporation MAB17822
Specifications
IGHA1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry (1:50-1:250) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
IGHA1 | |
Mouse | |
100 μg | |
Human, Monkey, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
7D5F12 | |
immunoglobulin heavy constant alpha 1 | |
In PBS (0.05% sodium azide) | |
FLJ14473, FLJ35065, FLJ35500, FLJ36402, FLJ39698, FLJ40001, FLJ41548, FLJ41552, FLJ41789, FLJ43248, FLJ43594, FLJ44293, FLJ46028, FLJ46621, FLJ46724, FLJ46811, FLJ46824, IgA1, MGC102857 | |
Recombinant protein corresponding to amino acids 207-353 of human IGHA1 from E. coli. | |
3493 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |