missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG3 Mouse, PE, Monoclonal, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$64.68 - $209.88

Specifications

Antigen IgG3
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5018663 Life Technologies
LS12474241
25 Tests Each for $64.68
5018664 Life Technologies
LS12474242
100 Tests Each for $209.88
Specifications

Description

The mouse IgG3, λ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG3
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
RUO
Conjugated
Monoclonal
10μL (0.1μg)/test