Please login to your online account to display your discounted pricing

IgG3 Mouse, Monoclonal, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$71.05 - $129.05

Specifications

Antigen IgG3
Host Species Murine
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunohistochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013054

 
life technologies
LS14474281
50μg Each for $71.05

5013055

 
life technologies
LS14474282
100μg Each for $129.05
Description & Specifications

Specifications

Antigen IgG3
Host Species Murine
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The mouse IgG3, λ is useful as an isotype control immunoglobulin.