Please login to your online account to display your discounted pricing

IgG3 Mouse, FITC, Monoclonal, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$114.55 - $187.05

Specifications

Antigen IgG3
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018627

 
life technologies
LS11474241
25 Tests Each for $114.55

5018628

 
life technologies
LS11474242
100 Tests Each for $187.05
Description & Specifications

Specifications

Antigen IgG3
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 10μL (1μg)/test

The mouse IgG3, λ is useful as an isotype control immunoglobulin.