Please login to your online account to display your discounted pricing

IgG2b Rat, Clone: eB149/10H5, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$114.55 - $259.55

Specifications

Antigen IgG2b
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124957

 
life technologies
LS69403180
25μg Each for $114.55

501124958

 
life technologies
LS69403182
100μg Each for $259.55
Description & Specifications

Specifications

Antigen IgG2b
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 506
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin.