missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2b Rat anti-Mouse, FITC, Clone: m2b-25G4, eBioscience™

Rat Monoclonal Antibody

$92.40 - $213.84

Specifications

Antigen IgG2b
Clone m2b-25G4
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry,Immunocytochemistry,Immunohistochemistry,Microscopy
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122119 Life Technologies
LS11422080
25μg Each for $92.40
501122120 Life Technologies
LS11422082
100μg Each for $213.84
Specifications

Description

The monoclonal antibody m2b-25G4 recognizes Mouse IgG2b antibodies and can be used as a second step reagent in flow cytometry and microscopy. This m2b-25G4 antibody does not recognize other mouse isotypes nor does it crossreact to rat IgG2b or any rat isotype antibodies.
Description & Specifications

Specifications

IgG2b
Rat
Flow Cytometry,Immunocytochemistry,Immunohistochemistry,Microscopy
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
m2b-25G4
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer