Please login to your online account to display your discounted pricing

IgG2b Mouse anti-Rat, eFluor 660, Clone: R2B-7C3, eBioscience™

Mouse Monoclonal Antibody

$101.50 - $249.40

Specifications

Antigen IgG2b
Clone R2B-7C3
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124370

 
life technologies
LS50481580
25μg Each for $101.50

501124371

 
life technologies
LS50481582
100μg Each for $249.40
Description & Specifications

Specifications

Antigen IgG2b
Clone R2B-7C3
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody R2B-7C3 recognizes Rat IgG2b antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2b or any mouse isotype antibodies.