Please login to your online account to display your discounted pricing

IgG2b, K Rat, PE-Cy5, Clone: eB149/10H5, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$230.55 - $361.05

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122927

 
life technologies
LS15403182
100μg Each for $317.55

5014103

 
life technologies
LS15403181
50μg Each for $230.55

5014105

 
life technologies
LS15403183
200μg Each for $361.05
Description & Specifications

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin.