Please login to your online account to display your discounted pricing

IgG2b, K Rat, FITC, Clone: eB149/10H5, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$114.55 - $317.55

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509805

 
affymetrix
11403185
500μg Each for $317.55

501129618

 
affymetrix
11403181
50μg Each for $114.55

501129627

 
affymetrix
11403182
100μg Each for $187.05
Description & Specifications

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The rat IgG2b, κ monoclonal antibody is useful as an isotype control immunoglobulin.