missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2b, K Rat, eFluor 660, Clone: eB149/10H5, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$117.48 - $302.28

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry,Immunohistochemistry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124351 Life Technologies
LS50403182
100μg Each for $302.28
5016443 Life Technologies
LS50403180
25μg Each for $117.48
Specifications

Description

The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG2b, K
Rat
RUO
Conjugated
Monoclonal
0.2mg/mL
eB149/10H5
Flow Cytometry,Immunohistochemistry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer