Please login to your online account to display your discounted pricing

IgG2b, K Rat, eFluor 570, Clone: eB149/10H5, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$131.95 - $249.40

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123605

 
life technologies
LS41403180
25μg Each for $131.95

501123606

 
life technologies
LS41403182
100μg Each for $249.40
Description & Specifications

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin.