missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2b, K Rat, Clone: eB149/10H5, Isotype Control, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$85.55 - $259.55

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014524 Life Technologies
LS16403181
50μg Each for $85.55
501129605 Life Technologies
LS16403185
500μg Each for $259.55
Specifications

Description

The rat IgG2b, κ monoclonal antibody is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG2b, K
Rat
RUO
Purified
Monoclonal
1mg/mL
eB149/10H5
Flow Cytometry
Unlabeled
Store at 2-8°C.
aqueous buffer, no sodium azide