missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2b, K Rat, APC, Clone: eB149/10H5, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$187.05 - $317.55

Specifications

Antigen IgG2b, K
Clone eB149/10H5
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015149 Life Technologies
LS17403181
50μg Each for $187.05
501128759 Life Technologies
LS17403182
100μg Each for $317.55
Specifications

Description

The rat IgG2b, κ monoclonal antibody is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG2b, K
Rat
RUO
Conjugated
Monoclonal
0.2mg/mL
eB149/10H5
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer