Please login to your online account to display your discounted pricing

IgG2b, K Mouse, PerCP-eFluor 710, Clone: eBMG2b, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$143.55 - $332.05

Specifications

Antigen IgG2b, K
Clone eBMG2b
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016050

 
affymetrix
46473280
25μg Each for $143.55

5016051

 
affymetrix
46473282
100μg Each for $332.05
Description & Specifications

Specifications

Antigen IgG2b, K
Clone eBMG2b
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal mouse IgG2b, κ is useful as an isotype control immunoglobulin.