Please login to your online account to display your discounted pricing

IgG2b, K Mouse, PE-eFluor 610, Monoclonal, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$143.55 - $361.05

Specifications

Antigen IgG2b, K
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124820

 
life technologies
LS61473280
25μg Each for $143.55

501124821

 
life technologies
LS61473282
100μg Each for $361.05
Description & Specifications

Specifications

Antigen IgG2b, K
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin.