Please login to your online account to display your discounted pricing

IgG2b, K Mouse, PE-Cy5, Clone: eBMG2b, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$129.05 - $216.05

Specifications

Antigen IgG2b, K
Clone eBMG2b
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122931

 
life technologies
LS15473242
100 Tests Each for $216.05

5014120

 
life technologies
LS15473241
25 Tests Each for $129.05
Description & Specifications

Specifications

Antigen IgG2b, K
Clone eBMG2b
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin.