Please login to your online account to display your discounted pricing

IgG2b, K Mouse, eFluor 570, Clone: eBMG2b, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$131.95 - $323.35

Specifications

Antigen IgG2b, K
Clone eBMG2b
Host Species Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123618

 
life technologies
LS41473280
25μg Each for $131.95

501123619

 
life technologies
LS41473282
100μg Each for $323.35
Description & Specifications

Specifications

Antigen IgG2b, K
Clone eBMG2b
Host Species Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin.