missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2b, K Mouse, eFluor 450, Clone: eBMG2b, Isotype Control, eBioscience™

Mouse Monoclonal Isotype Control

$130.68 - $262.68

Specifications

Antigen IgG2b, K
Clone eBMG2b
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016355 Life Technologies
LS48473280
25μg Each for $130.68
5016356 Life Technologies
LS48473282
100μg Each for $262.68
Specifications

Description

The monoclonal mouse IgG2b, κ is useful as an isotype control immunoglobulin.
Description & Specifications

Specifications

IgG2b, K
Murine
RUO
Conjugated
Monoclonal
0.2mg/mL
eBMG2b
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer