missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IgG2b kappa, APC, clone: eBMG2b, Isotype Control, eBioscience™

Mouse Isotype Control

Manufacturer:  Life Technologies LS17473242

 View more versions of this product

Catalog No. 501129582



Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin.Applications Reported: APC Mouse IgG2b Iso Cntrl has been reported for use in flow cytometric analysis.Applications Tested: APC Mouse IgG2b Isotype Control is offered in 2 formats: - μg size: has been tested by flow cytometric analysis of normal human peripheral blood cells. This should be used at the same concentration as the experimental antibody. - test size: has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at test size: 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Mouse IgG2b kappa
Control
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
IgG2, IgG2b, IgG2bk
IgG2b, kappa, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Control, Flow Cytometry
eBMG2b
APC
Liquid
Murine
Affinity chromatography
RUO
Isotype Control
Not Applicable
Documents

For Research Use Only.