Please login to your online account to display your discounted pricing

IgG2a Rat, Clone: eBR2a, Isotype Control, eBioscience™

Rat Monoclonal Isotype Control

$114.55 - $288.55

Specifications

Antigen IgG2a
Clone eBR2a
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124959

 
life technologies
LS69432180
25μg Each for $114.55

501124960

 
life technologies
LS69432182
100μg Each for $288.55
Description & Specifications

Specifications

Antigen IgG2a
Clone eBR2a
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 506
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal rat IgG2a is useful as an isotype control immunoglobulin.