Please login to your online account to display your discounted pricing

IgG2a Rat anti-Mouse, eFluor 615, Clone: m2a-15F8, eBioscience™

Rat Monoclonal Antibody

$160.95 - $397.30

Specifications

Antigen IgG2a
Clone m2a-15F8
Host Species Rat
Species Reactivity Mouse
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123670

 
life technologies
LS42421080
25μg Each for $160.95

501123671

 
life technologies
LS42421082
100μg Each for $397.30
Description & Specifications

Specifications

Antigen IgG2a
Clone m2a-15F8
Host Species Rat
Species Reactivity Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate eFluor 615
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies.