missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2a Rat anti-Mouse, eFluor 570, Clone: m2a-15F8, eBioscience™

Rat Monoclonal Antibody

$146.52 - $361.68

Specifications

Antigen IgG2a
Clone m2a-15F8
Host Species Rat
Species Reactivity Mouse
Applications Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123607 Life Technologies
LS41421080
25μg Each for $146.52
501123608 Life Technologies
LS41421082
100μg Each for $361.68
Specifications

Description

The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies.
Description & Specifications

Specifications

IgG2a
Rat
Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
eFluor 570
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
m2a-15F8
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer