missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgG2a Rat anti-Mouse, Biotin, Clone: m2a-15F8, eBioscience™

Rat Monoclonal Antibody

$79.20 - $187.44

Specifications

Antigen IgG2a
Clone m2a-15F8
Host Species Rat
Applications Immunocytochemistry,Immunohistochemistry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122543 Life Technologies
LS13421082
100μg Each for $187.44
501128599 Life Technologies
LS13421080
25μg Each for $79.20
Specifications

Description

The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies.
Description & Specifications

Specifications

IgG2a
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
m2a-15F8
Immunocytochemistry,Immunohistochemistry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL